Diversified Biotech Synergel™ Agarose Additive
Catalogue # 52857-970
Fournisseur: Diversified Biotech
Small Business Enterprise
Spécifications
- Description:Synergel Agarose Additive
- Cat. no.:52857-970
- Supplier No.:SYN100
Spécifications
À propos de cet article
Better sieving than agarose, 1 gram Synergel = 2 grams agarose.High Resolution
Synergel/Agarose gels exhibit improved spatial separation and tighter banding for DNA fragments of 50-2000bp. No Background Fluorescence. The superior optical clarity permits high quality photo-documentation. Increased Load Capacity—Accommodates greater quantities of DNA reducing band tailing and smearing. Easy To Use; mix, boil, and pour.