3B Scientific® Skull With 5 Part Brain
# de catalogue 470118-916
Fournisseur: American 3B Scientific
Spécifications
- Description:Skull With 5 Part Brain
- Height:14 cm
- Cat. no.:470118-916
- Length:18 cm
- Width:12 cm
- Supplier No.:A20/9
- No. of Parts:8
Spécifications
A propos de cet article
The 3 part medical quality human skull replica is a first choice for basic anatomical studies of the skull. This skull model includes an anatomically correct 5 part brain.
- 3 year manufacturers warranty
- Includes 3B smart anatomy
The brain is midsagitally divided human brain replica and is cast from an original specimen, lending anatomical accuracy to this model.
The components of the left brain half are: Frontal and parietal lobe, Temporal and occipital lobe, Encephalic trunk, and Cerebellum.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought