HyperSep Universal Vacuum Manifold, Thermo Scientific
# de catalogue 10047-012
Fournisseur: Thermo Scientific
Spécifications
- Description:Universal Vacuum Manifold
- Cat. no.:10047-012
- Supplier No.:60104-230
Spécifications
A propos de cet article
Allows processing of SPE and SLE cartridges (3,4 flangeless and 6 mL) or 96-well plates
Allows flexibility in sample processing.
Delivery: Includes manifold, base/gauge, flask and stopper, tubing and spigots.
Customers Who Bought This Also Bought