HyperSep 96-Well Plate Manifold, Thermo Scientific
# de catalogue 10047-010
Fournisseur: Thermo Scientific
Spécifications
- Description:Vacuum Manifold, 96-Well Plate
- Cat. no.:10047-010
- Supplier No.:60103-351
Spécifications
A propos de cet article
Provides processing of 96-well SPE, SLE, Protein precipitation and Lab/Filter plates
This format provides higher throughput processing of samples allowing for higher efficency workflows and reproducible results.
Delivery: 96-Well Plate Vacuum Manifold includes base, lid, waist collection tray and vacuum gauge.
Customers Who Bought This Also Bought