Economy Molded Cylinders, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
These cylinders feature permanently molded graduations that will not fade after repeated autoclaving.
- Durable molded graduations
- Shatter resistant - reduces breakage and injury
- Chemically resistant to most acids, bases and many common solvents
- Autoclavable
- Choose glass-like clarity of polymethylpentene (PMP) or
- Choose economical translucent polypropylene (PP)
- Food grade product
Meets stringent international quality criteria according to ISO 6706 standards. Tolerances according to DIN 12 680 TT, Class B. Maximum temperature: PP 121°C, PMP 135°C.
Spécifications
- Catalog No:
- 30620-304
- 13450-664
- 13450-662
- 13450-670
- 13450-646
- 13450-660
- 30620-288
- 13450-658
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall