Accucore™ Phenyl-Hexyl Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Separate aromatic and moderately polar compounds with improved speed and resolution using Thermo Scientific™ Accucore Phenyl-Hexyl columns.
Rugged 2.6 µm solid core particles enable fast, high efficiency separations at low backpressures. The phenyl-hexyl stationary phase offers a mixed-mode separation mechanism: the C6 chain provides hydrophobic retention while the phenyl ring enhances pi-pi interactions. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10038-712
- 10038-714
- 10038-716
- 10038-740
- 10038-742
- 10039-098
- 10038-704
- 10039-100
- 10039-102
- 10038-706
- 10038-708
- 10038-710
- Description:
- 10038-712
- 10038-714
- 10038-716
- 10038-740
- 10038-742
- 10039-098
- 10038-704
- 10039-100
- 10039-102
- 10038-706
- 10038-708
- 10038-710
- Carbon load:
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- pH range:
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- Surface area:
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g