Accucore™ C8 Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
À propos de cet article
Improve separation speed and resolution while maintaining low backpressures using Thermo Scientific™ Accucore C8 columns.
Rugged 2.6 µm solid core particles ensure high efficiencies and enable compatibility with both HPLC and UHPLC platforms. The high coverage C8 bonded phase provides optimal retention of analytes with moderate hydrophobicity. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10038-816
- 10038-838
- 10038-818
- 10038-840
- 10038-820
- 10038-822
- 10038-828
- 10038-830
- 10038-834
- 10038-812
- 10038-814
- 10038-836
- Description:
- 10038-816
- 10038-838
- 10038-818
- 10038-840
- 10038-820
- 10038-822
- 10038-828
- 10038-830
- 10038-834
- 10038-812
- 10038-814
- 10038-836
- Carbon load:
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- 5%
- pH range:
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- Surface area:
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g