Accucore™ C18 Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Achieve fast, high-resolution separations at low backpressures using Thermo Scientific™ Accucore C18 columns.
Rugged 2.6 µm solid core particles ensure high efficiencies and enable compatibility with both HPLC and UHPLC platforms. The high bonded phase coverage provides optimal retention of a broad range of non-polar analytes across multiple applications. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10038-842
- 10038-844
- 10038-848
- 10038-872
- 10038-850
- 10038-874
- 10038-852
- 10038-879
- 10038-870
- 10038-876
- 10038-856
- 10038-858
- 10038-860
- 10038-862
- 10038-866
- 10038-868
- Carbon load:
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- 9%
- pH range:
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- Surface area:
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g