Accucore™ RP-MS Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
À propos de cet article
Optimize LC/MS analyses with Thermo Scientific™ Accucore RP-MS columns.
Rugged 2.6 µm solid-core particles enable fast, high-efficiency separations at low backpressures. The stationary phase provides slightly lower retention than C18 and has more effective surface coverage for improved peak efficiencies. Robust bonding technology and automated packing procedures ensure excellent reproducibility and long column lifetimes.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10039-112
- 10039-134
- 10039-114
- 10039-116
- 10039-118
- 10039-120
- 10039-126
- 10039-104
- 10039-128
- 10039-106
- 10039-108
- 10039-130
- 10039-110
- Description:
- 10039-112
- 10039-134
- 10039-114
- 10039-116
- 10039-118
- 10039-120
- 10039-126
- 10039-104
- 10039-128
- 10039-106
- 10039-108
- 10039-130
- 10039-110
- Carbon load:
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- 7%
- pH range:
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- Surface area:
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g
- 130 m²/g