Hypersil GOLD™ Phenyl HPLC Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Get excellent retention and unique selectivity for aromatic and moderately polar analytes with Thermo Scientific™ Hypersil GOLD™ Phenyl reversed phase HPLC columns.
A butyl (C4) linker in the phenyl bonded phase allows for superior alignment of the phenyl ring with aromatic molecules, thus enhancing pi-pi interactions and improving the retention of the molecules. The C4 linker also provides the stationary phase with moderate hydrophobicity, making it ideal for the separation of analyte mixtures with varying polarity and aromaticity. The outstanding peak shape results in greater sensitivity.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10041-766
- 10045-730
- 10045-762
- 10041-770
- 10045-746
- 10041-780
- 10041-778
- 10041-776
- 10045-734
- 10041-788
- 10041-792
- 76332-252
- 10045-756
- 10041-758
- Description:
- 10041-766
- 10045-730
- 10045-762
- 10041-770
- 10045-746
- 10041-780
- 10041-778
- 10041-776
- 10045-734
- 10041-788
- 10041-792
- 76332-252
- 10045-756
- 10041-758
- Carbon load:
- 8.50%
- 8.50%
- 8.50%
- 8.50%
- 8.50%
- 8.50%
- 8.50%
- 8.50%
- 8.50%
- 8.50%
- 8.50%
- 8.0%
- 8.50%
- 8.50%
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g