ProSwift RP-4H Protein Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Run protein separations at exceptionally high flow rates with Thermo Scientific™ ProSwift RP columns.
These reversed-phase monolith columns deliver the outstanding resolving power of nonporous analytical media combined with faster separations and analysis than other bead-based columns. ProSwift polymer monolith columns achieve long column lifetime, exhibit reproducibility even after hundreds of runs, and have broad pH stability, providing high stringent wash capability (1 M NaOH).
Spécifications
- Catalog No:
- 10044-582
- 10044-616
- Description:
- 10044-582
- 10044-616
- pH range:
- 1 to 14
- 1 to 14