VWR® Cleanroom Paper for Dot Matrix Applications
# de catalogue 10027-958
Fournisseur: VWR International
Spécifications
- Description:Continuous cleanroom paper
- Cat. no.:10027-958
- Dimensions:9.5×11"
Spécifications
A propos de cet article
Ideal for dot matrix printer applications in critical environments, where very low particle and ESD generation is important.
- Low particle generation
2,300 sheets per case.
Customers Who Bought This Also Bought