VWR® Cleaning Cloths
# de catalogue 21902-990
Fournisseur: VWR International
Spécifications
- Description:Cleaning Cloths
- L×W:8" x 8"
- Cat. no.:21902-990
Spécifications
A propos de cet article
This non-woven material is a unique, high-quality fabric made specifically for the Graphic Arts Industry.
- Soft
- Economical
- Absorbent
- Non-scratching
This 100% cotton cloth wipe will not scratch plates or proofs. These wipes are dispensed from a roll of 100, 8×8" wipes per roll. They are perfect for cleaning any lenses or computer screens. Some other great uses are blending pastels, or cleaning graphic arts equipment.
Customers Who Bought This Also Bought