VWR® Rubber Tubing, Natural Red
Fournisseur: VWR International
Certificats
A propos de cet article
High-quality, durable, natural rubber tubing for general laboratory use.
- Durometer hardness: Shore A, 50
- Tensile strength: 2500-3500 psi
Seamless, with uniform medium wall and low sulfur content.
Customers Who Bought This Also Bought