Hypersil GOLD™ HILIC HPLC Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Thermo Scientific™ Hypersil GOLD™ HILIC (hydrophilic interaction liquid chromatography) HPLC columns.
These columns deliver straightforward separation without the need for sample derivatization or ion-pair reagents. Featuring outstanding peak shape and selectivity, the columns provide improved sensitivity for MS detection. With 1.9, 3, and 5 µm particle sizes, Hypersil GOLD HILIC columns are ideal for applications within drug discovery, food testing, and environmental analysis.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10045-792
- 10041-942
- 10045-794
- 10041-938
- 10041-948
- 10045-790
- 10041-944
- 10041-952
- 10045-802
- 10045-804
- 10041-956
- 10041-964
- 10041-916
- 10041-928
- 10041-924
- 10045-810
- 10045-812
- 10041-922
- 10041-930
- Description:
- 10045-792
- 10041-942
- 10045-794
- 10041-938
- 10041-948
- 10045-790
- 10041-944
- 10041-952
- 10045-802
- 10045-804
- 10041-956
- 10041-964
- 10041-916
- 10041-928
- 10041-924
- 10045-810
- 10045-812
- 10041-922
- 10041-930
- Carbon load:
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- 6%
- pH range:
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g