Hypersil GOLD™ Silica HPLC Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Achieve outstanding peak shape and sensitivity with Thermo Scientific™ Hypersil GOLD™ Silica HPLC columns.
The unbonded, highly pure, base-deactivated silica media efficiently separates non-polar and moderately polar organic compounds by normal phase chromatography. Available 1.9 µm particle size columns can improve the speed and efficiency of your applications.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10041-128
- 10041-126
- 10041-122
- 10041-154
- 10041-184
- 10041-192
- 10041-190
- 10041-118
- 10041-136
- 10041-158
- Description:
- 10041-128
- 10041-126
- 10041-122
- 10041-154
- 10041-184
- 10041-192
- 10041-190
- 10041-118
- 10041-136
- 10041-158
- pH range:
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g