Hypersil GOLD™ Amino HPLC Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Achieve excellent chromatographic separations in weak anion exchange, reversed phase, and normal phase modes with Thermo Scientific™ Hypersil GOLD™ Amino HPLC columns.
These columns, featuring a high-performance aminopropyl phase, provide outstanding peak shape and sensitivity and are available with a 1.9 μm particle size for high speed, high efficiency separations.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10041-656
- 10041-620
- 10041-684
- 10041-682
- 10041-648
- 10041-646
- 10041-602
- 10041-644
- 10041-622
- 10041-614
- 10041-612
- 10041-674
- 10041-608
- Description:
- 10041-656
- 10041-620
- 10041-684
- 10041-682
- 10041-648
- 10041-646
- 10041-602
- 10041-644
- 10041-622
- 10041-614
- 10041-612
- 10041-674
- 10041-608
- Carbon load:
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- pH range:
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g