Hypersil GOLD™ CN HPLC Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
For alternative selectivity in both reversed phase and normal phase chromatography, use Thermo Scientific™ Hypersil GOLD™ CN HPLC columns.
Based on highly pure silica, these columns offer excellent peak shapes and outstanding sensitivity with less retention for faster analysis. Analyze a wide range of compounds, from steroids and polyphenols in reversed phase mode to surfactants and other polar species in normal phase mode.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10041-744
- 10045-706
- 10041-742
- 10045-670
- 10045-674
- 10045-684
- 10045-718
- 10041-730
- 10045-678
- 10045-680
- 10041-690
- 10045-694
- Description:
- 10041-744
- 10045-706
- 10041-742
- 10045-670
- 10045-674
- 10045-684
- 10045-718
- 10041-730
- 10045-678
- 10045-680
- 10041-690
- 10045-694
- Carbon load:
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- 4%
- pH range:
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g