MAbPac™ Protein A Affinity Columns, Thermo Scientific
# de catalogue 10044-912
Fournisseur: Thermo Scientific
Spécifications
- Column Format:Analytical
- I.D.:4.00 mm
- Length:35 mm
- Particle size:12 µm
- pH range:2.5 to 7.5
- Description:10044-912
- Bed Weight:100 µm
- Cat. no.:10044-912
- Packing Material:Divinylbenzene, IgG
- Phase:Protein A
- Supplier No.:082539
Spécifications
A propos de cet article
Run fast, accurate titer analysis of monoclonal antibodies (MAbs) in harvest cell cultures with the Thermo Scientific™ MAbPac Protein A column.
- This affinity chromatography column provides high throughput and accurate analyses through a combination of low backpressure and high efficiency
- The unique column format makes automation of loading, binding, elution, and collection using biocompatible systems quick and easy
A non-porous, polymeric resin with a hydrophobic divinylbenzene core and a hydrophilic surface optimized for affinity separation makes this column rugged and reproducible.
Customers Who Bought This Also Bought