ProPac™ HIC Hydrophobic Interaction Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Achieve high-resolution separation of proteins and peptides under non-denaturing conditions with Thermo Scientific™ ProPac HIC-10 hydrophobic interaction chromatography columns.
Developed with multi-functional ligand attachments, these silica-based columns provide exceptional hydrolytic stability under the highly aqueous conditions used in HIC.
Spécifications
- Catalog No:
- 10040-598
- 10044-518
- 10044-520
- 10044-522
- Max. pressure:
- 3500 psi
- 3500 psi
- 3500 psi
- 3500 psi
- pH range:
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- Surface area:
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- Temperature limit:
- 40 °C
- 40 °C
- 40 °C
- 40 °C