ProPac™ WAX-10 Anion Exchange Protein Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
For exceptionally high resolution and efficiency anion-exchange separation of protein variants, use Thermo Scientific™ ProPac SAX and WAX strong and weak anion-exchange columns.
- Based on a nonporous 10 µm core particle that provides pressure stability, these columns can resolve isoforms that differ by a single charged residue
A thin, hydrophilic layer prevents unwanted secondary interactions. The grafted anion-exchange surface provides pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity.
Spécifications
- Catalog No:
- 10044-506
- 10044-538
- 10044-332
- 10044-390
- 10044-392
- 10044-504
- Max. pressure:
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- pH range:
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- Temperature limit:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C