ProPac™ SCX Cation Exchange Protein Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
For separations of proteins and their variants using Thermo Scientific™ ProPac SCX and WCX strong and weak cation-exchange columns, which provide exceptionally high resolution.
Based on stationary phases composed of 10 µm nonporous polymeric beads, these columns resolve isoforms that differ by a single charged residue. A thin, hydrophilic layer grafted to the particle core eliminates unwanted secondary interactions. The cation-exchange surface provides pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity.
Spécifications
- Catalog No:
- 10043-310
- 10044-384
- 10044-532
- 10044-500
- 10044-502
- 10044-328
- 10042-826
- Description:
- Column
- Column
- Column
- Column
- Column
- Column
- Column
- pH range:
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12