ProPac™ WCX-10 Cation Exchange Protein Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
For separations of proteins and their variants using Thermo Scientific™ ProPac SCX and WCX strong and weak cation-exchange columns, which provide exceptionally high resolution.
Based on stationary phases composed of 10 µm nonporous polymeric beads, these columns resolve isoforms that differ by a single charged residue. A thin, hydrophilic layer grafted to the particle core eliminates unwanted secondary interactions. The cation-exchange surface provides pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity.
Spécifications
- Catalog No:
- 10044-322
- 10044-508
- 10044-510
- 10042-822
- 10044-512
- 10044-380
- 10044-382
- 10044-346
- 10044-324
- 10044-326
- 10044-348
- Max. pressure:
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- 3000 psi
- pH range:
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- Temperature limit:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C