Hypersil GOLD™ aQ HPLC Columns, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Hypersil GOLD™ aQ is a polar endcapped C18 phase, which offers superior retention of polar compounds.
- Ensures optimized peak shape, resolution, sensitivity and efficiency
- Provides only controlled secondary interactions to ensure excellent peak shape for all analyte types
- Polar endcapping of media also makes it usable in 100% aqueous mobile phases without the risk of loss of performance or poor stability
The additional interaction mechanism often provides selectivity differences over the traditional alkyl chain chemistries, and offers a solution for the separation of polar compounds which exhibit insufficient retention on pure alkyl chain phases under typical reversed phase mobile phase conditions.
Specifications
- Catalog No:
- 10041-348
- 10045-548
- 10041-346
- 10045-550
- 10041-344
- 10041-342
- 10041-378
- 10041-420
- 10045-542
- 10045-544
- 10041-416
- 10045-502
- 10041-430
- 10041-394
- 10045-516
- 10041-436
- 10045-530
- 10045-532
- 10041-334
- 10041-398
- 10041-330
- 10041-374
- 10041-328
- 10041-446
- 10041-414
- 10045-536
- 10041-340
- Description:
- 10041-348
- 10045-548
- 10041-346
- 10045-550
- 10041-344
- 10041-342
- 10041-378
- 10041-420
- 10045-542
- 10045-544
- 10041-416
- 10045-502
- 10041-430
- 10041-394
- 10045-516
- 10041-436
- 10045-530
- 10045-532
- 10041-334
- 10041-398
- 10041-330
- 10041-374
- 10041-328
- 10041-446
- 10041-414
- 10045-536
- 10041-340
- Carbon load:
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- pH range:
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g