Hypersil GOLD™ C8 HPLC Columns, Thermo Scientific
Fournisseur: Avantor
Certificats
A propos de cet article
For analytes with medium hydrophobicity or when a less hydrophobic phase is required to obtain optimum retention, use Thermo Scientific™ Hypersil GOLD™ C8 HPLC columns for outstanding sensitivity and excellent peak shape.
The selectivity is similar to C18 columns with a predictable elution order but less retention. The lower hydrophobicity allows compounds to elute more quickly, yielding faster separations.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10041-258
- 10041-256
- 10041-266
- 10041-262
- 10041-270
- 10045-482
- 10041-280
- 10045-484
- 10045-488
- 10041-276
- 10041-202
- 10041-290
- 10041-284
- 10045-492
- 10041-212
- 76332-372
- 10041-296
- 10041-304
- 10041-216
- 10041-298
- 10041-312
- 10041-222
- 10041-310
- 10041-220
- 10041-228
- 10041-226
- 10041-246
- 10041-242
- 10041-240
- 10041-254
- 10041-252
- 10041-248
- Description:
- 10041-258
- 10041-256
- 10041-266
- 10041-262
- 10041-270
- 10045-482
- 10041-280
- 10045-484
- 10045-488
- 10041-276
- 10041-202
- 10041-290
- 10041-284
- 10045-492
- 10041-212
- 76332-372
- 10041-296
- 10041-304
- 10041-216
- 10041-298
- 10041-312
- 10041-222
- 10041-310
- 10041-220
- 10041-228
- 10041-226
- 10041-246
- 10041-242
- 10041-240
- 10041-254
- 10041-252
- 10041-248
- Carbon load:
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- pH range:
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 - 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g