Hypersil GOLD™ HPLC Cartridges and Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Achieve exceptional peak shape and resolution for your HPLC and LC/MS applications with Thermo Scientific Hypersil GOLD HPLC columns.
- Endcapped, ultrapure, silica-based columns
- Excellent resolution, efficiency, and sensitivity
- Accuracy and quality of your analytical data
These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. With their excellent resolution, efficiency, and sensitivity, Hypersil GOLD columns give you confidence in the accuracy and quality of your analytical data. 1.9 µm particle size columns can improve the speed and efficiency of your applications.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder, 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder.
Spécifications
- Catalog No:
- 10041-020
- 10045-230
- 10045-328
- 10045-332
- 10045-238
- 10045-334
- 10045-336
- 10045-340
- 10045-342
- 10045-256
- 10045-274
- 10045-276
- 10045-278
- 10041-000
- 10045-280
- 10045-282
- Carbon load:
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- pH range:
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- 1 to 11
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g