Accucore™ 150-Amide-HILIC Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Achieve separation of hydrophilic biomolecules using Thermo Scientific Accucore 150-Amide-HILIC columns.
Combined with the larger pore size of the solid core particles, Accucore 150-Amide-HILIC is well suited for separating a carbohydrates and peptides.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10038-892
- 10038-894
- 10038-898
- 10038-900
- 10038-880
- 10038-902
- 10038-882
- 10038-884
- 10038-886
- 10038-888
- 10038-890
- Description:
- Column
- Column
- Column
- Column
- Column
- Guard Cartridge
- nanoViper Column
- Column
- Column
- Column
- Column
- pH range:
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- Surface area:
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g