Accucore™ 150-C4 Columns, Thermo Scientific
Fournisseur: Avantor
Certificats
A propos de cet article
The 150A pore diameter solid core particles used in Thermo Scientific Accucore 150-C4 columns are designed specifically to provide the optimum combination of retention and resolution for proteins and larger peptides.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Spécifications
- Catalog No:
- 10039-174
- 10039-176
- 10039-178
- 10039-156
- 10039-158
- 10039-180
- 10039-160
- 10039-162
- 10039-170
- 10038-906
- 10038-908
- Description:
- Column
- Column
- Column
- Column
- Column
- Column
- Column
- Column
- Column
- Column
- nanoViper Column
- Carbon load:
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- 2%
- pH range:
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- 2 to 9
- Surface area:
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g
- 80 m²/g