A propos de cet article
Tygon S3™ B-44-3 Beverage Tubing from Saint-Gobain Performance Plastics is now phthalate-free.
Tygon S3™ B-44-3 Beverage Tubing is as clear as glass for easy visual monitoring of flow. Lightweight and flexible for easy and quick installation. Broad chemical resistance. Non-wetting properties allow easy cleaning and complete drainage.
Saint-Gobain is proud to be among the first companies to offer sustainable flexible tubing products. This bio-based tubing line combines the high performance standards customers demand with an eco-friendly design. Versatile, high-performance Tygon S3™ B-44-3 Beverage tubing is specifically formulated for transferring a wide variety of beverages, including soft drinks, fruit juices, flavored teas, and bottled water. In virtually all cases, Tygon S3™ B-44-3 tubing will not affect the taste or odor of products transferred through it, while its excellent non-wetting properties facilitate complete drainage and permit simple flush-cleaning. Many of the unique properties inherent to this tubing line also apply to a wide variety of complex applications, ranging from fine cosmetic production to the dispensing of water-based printer inks found in the publishing industry. This versatility and proven performance have made it today's most widely specified clear, flexible, plastic tubing. Light in weight and easy to handle, Tygon S3™ B-44-3 tubing can be put into service quickly. It readily bends to accommodate abrupt corners and obstructions, requiring a minimum of couplings and fittings. Its flexibility can save up to one-third the footage and much of the labor required to install rigid stainless steel or plastic piping. The glass-like clarity ensures visual monitoring of flow is easy. This is particularly helpful in controlling or adjusting flow during processing.
Spécifications
- Catalog No:
- 89404-066
- 89404-068
- 89404-092
- 89404-094
- 89404-096
- 89404-098
- 89404-100
- 89404-104
- 89404-106
- 89404-110
- 89404-114
- 63010-960
- 89404-154
- 89404-070
- 89404-072
- 89404-074
- 89404-076
- 89404-080
- 89404-082
- 89404-084
- 89404-088
- 89404-130
- 89404-116
- 89404-102
- 89404-124
- 89404-120
- 89404-148
- 89404-144
- Model:
- 89404-066
- 89404-068
- 89404-092
- 89404-094
- 89404-096
- 89404-098
- 89404-100
- 89404-104
- 89404-106
- 89404-110
- 89404-114
- 63010-960
- 89404-154
- 89404-070
- 89404-072
- 89404-074
- 89404-076
- 89404-080
- 89404-082
- 89404-084
- 89404-088
- 89404-130
- 89404-116
- 89404-102
- 89404-124
- 89404-120
- 89404-148
- 89404-144
- Max. vacuum:
- 29.9"
- 29.9"
- 29.9"
- 29.9"
- 16.0"
- 29.9"
- 29.9"
- 25.0"
- 29.9"
- 6.0"
- 25.0"
- 29.9"
- 29.9"
- 29.9"
- 25.0"
- 29.9"
- 16.0"
- 11.0"
- 29.9"
- 29.9"
- 6.0"
- 4.0"
- 16.0"
- 11.0"
- 6.0"
- 11.0"
- 29.9"
- 29.9"
- Working pressure:
- 100.0 psi
- 43.0 psi
- 47.0 psi
- 60.0 psi
- 28.0 psi
- 40.0 psi
- 50.0 psi
- 34.0 psi
- 44.0 psi
- 19.0 psi
- 34.0 psi
- 43.0 psi
- 70.0 psi
- 74.0 psi
- 34.0 psi
- 60.0 psi
- 28.0 psi
- 25.0 psi
- 43.0 psi
- 60.0 psi
- 19.0 psi
- 16.0 psi
- 29.0 psi
- 25.0 psi
- 20.0 psi
- 25.0 psi
- 60.0 psi
- 60.0 psi