SilTite Capillary Column Connectors, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
Certificats
A propos de cet article
For use with capillary GC columns.
Customers Who Bought This Also Bought
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$343.64
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$121.04
WHEATON® MicroLiter Crimp-Top Vials, 11 mm,Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$79.72