GFM Pro Electronic Flowmeter, Thermo Scientific | Avantor
GFM Pro Electronic Flowmeter, Thermo Scientific
GFM Pro Flowmeter
Catalog # 10057-406
Fournisseur #:  66002-010
GFM Pro Electronic Flowmeter, Thermo Scientific
Catalog # 10057-406
Supplier:  
GFM Pro Electronic Flowmeter, Thermo Scientific
GFM Pro Electronic Flowmeter, Thermo Scientific
GFM Pro Flowmeter
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
RESTEK PROFLOW 6000 Catalog #89205-666
Le produit 89205-666 est un remplacement de 10057-406 et a le même emballage et les mêmes spécifications, mais peut être d'une quantité différente.
Product Details & Documents
The Thermo Scientific GFM Pro Flowmeter is specifically designed for use with gas chromatography instruments.
  • Portable unit and can be hand-held
  • Has optional stand for bench-top convenience
This versatile product is an electronic device capable of measuring volumetric flow for all types of gases. Real-time measurements can be made for various types of flow paths including continually changing gas types.
Spécifications
  • Désignation:
    GFM Pro Flowmeter