Hypersil GOLD™ PFP HPLC Columns, Thermo Scientific
Fournisseur: Avantor
Certificats
A propos de cet article
Analyze difficult-to-resolve mixtures of halogenated compounds and non-halogenated polar compounds with Thermo Scientific™ Hypersil GOLD™ PFP (pentafluorophenyl) HPLC columns.
These columns provide an alternative selectivity to C18 columns in reversed phase applications with excellent peak shape and sensitivity.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm ID require the 1 mm UNIGUARD Holder (10046-746), 2.1/3 mm ID require the 2/3 mm UNIGUARD Holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD Holder (10046-744)
Spécifications
- Catalog No:
- 10045-616
- 10045-622
- 10041-516
- 10041-522
- 10041-454
- 10045-576
- 10045-578
- 10045-580
- 10041-538
- 10045-574
- 10041-464
- 10045-588
- 10041-548
- 10041-474
- 10041-470
- 10041-468
- 10045-604
- 10041-486
- 10045-610
- Description:
- 10045-616
- 10045-622
- 10041-516
- 10041-522
- 10041-454
- 10045-576
- 10045-578
- 10045-580
- 10041-538
- 10045-574
- 10041-464
- 10045-588
- 10041-548
- 10041-474
- 10041-470
- 10041-468
- 10045-604
- 10041-486
- 10045-610
- Carbon load:
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- pH range:
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g