Retention Gap Kits, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
Certificats
A propos de cet article
Support Thermo Scientific gas chromatography consumables.
Delivery: Kits contain: Length of deactivated fused silica tubing, 1 Mini-union, 5 ferrules.
Customers Who Bought This Also Bought
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
81,56 $ to 628,06 $
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
184,38 $
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
131,04 $
SureSTART™ Glass Screw Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
182,38 $ to 196,03 $
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
461,12 $ to 461,12 $
SureSTART™ Caps for 20 ml Glass Crimp Top Headspace Vials, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
1 418,93 $ to 1 418,93 $