GC Installation Kit, Thermo Scientific
# de catalogue 10057-430
Fournisseur: Thermo Scientific
Spécifications
- Description:GC Installation Kits
- Cat. no.:10057-430
- Supplier No.:60180-888
Spécifications
A propos de cet article
This installation kit includes: Tubing cutter, 1/8 x 1/4" reamer, 7/16in. wrench, 1/2in. wrench, 1/8in. brass tees (4), 1/8in. brass nuts (10), Brass front and back ferrules (10), 15.2m instrument grade cleaned 1/8in. copper tubing.
Customers Who Bought This Also Bought