Reacti-Vap Evaporators, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Manifolds for easy sample evaporation, with integrated pressure-relief valve to protect against excessive gas flow and dangerous pressure build up
Easy set up: attach to corresponding Reacti-Therm Module, attach tubing from gas supply, and lower into position over samples and start gas flow.
Ordering information: For a list of compatible aluminum alloy blocks, please see Reacti-Block Aluminum Blocks, Thermo Scientific.
Customers Who Bought This Also Bought