Reacti-Therm Heating and Stirring Modules, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
Certificats
A propos de cet article
Reliable and easy-to-use for constant temperatures.
Customers Who Bought This Also Bought
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
81,80 $
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
461,12 $ to 461,12 $
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
152,32 $
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
79,21 $ to 82,14 $
SureSTART™ Polypropylene Screw Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
38,19 $ to 40,16 $
SureSTART™ Glass Screw Top Vials, 2 ml, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
31,00 $