Gold Cross Inlet Base Seals for Agilent GCs, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Gold plating provides a surface with exceptional inertness suitable for analysis of highly active compounds.
Customers Who Bought This Also Bought