Gold Inlet Base Seals for Agilent GCs, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Optimize GC analysis of highly active compounds.
- Precision-machined from high-grade stainless steel
- Provides an excellent seal
They are gold plated to ensure a chemically inert surface for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs), and phenols. These inlet base seals are compatible with Thermo Scientific™ TRACE™ 1300 Series GC split and splitless injection ports and with all Agilent™ GC 5890/6890/7890 split and splitless injection ports.
Customers Who Bought This Also Bought