Thermo Scientific Super Clean Cartridge Filter Baseplates, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
A range of baseplate options allows configuration to your individual requirements. Needle valves ensure that the gas line is not contaminated during cartridge change.
Baseplates require one-time installation only.
Delivery: Super Clean Cartridge Filters are not included.
Customers Who Bought This Also Bought