ProPac™ PA1 Protein and Peptide Columns, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Efficiently analyze and purify hydrophilic anionic proteins and peptides with the Thermo Scientific™ ProPac PA1 Protein and Peptide column.
These strong anion-exchange columns provide high-resolution separations of proteins with pI values from 3 to 11.
Spécifications
- Catalog No:
- 10040-512
- 10040-510
- 10045-854
- Description:
- 10040-512
- 10040-510
- 10045-854
- pH range:
- 3 to 11
- 3 to 11
- 3 to 11