Mininert Valves, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Thermo Scientific Mininert Valves are screw caps that have integrated resealable valves to allow repeated and unlimited syring needle access to samples
Slide the valve one way to open the needle-port. Slide the valve back to close and completely seal the close
Customers Who Bought This Also Bought