PTFE Rubber Laminated Discs, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Provides a highy inert and unreactive seal.
- Rigid and more difficult to puncture
- Disc thickness of 60 mils
Made of white pharmaceutical rubber with 5 mils of PTFE coating bonded to one side, they are excellent for use as cap liners when a highly inert nonreactive seal is desired. Discs are autoclavable with no loss of integrity after heating above 100 °C for five hours.
Customers Who Bought This Also Bought