SP Bel-Art Octagon Spinbar Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape.
- PTFE low-friction coating
- Durable and inert
- Interrupted profile provides greater surface area and added turbulence
- Pivot ring aids in reducing friction and chattering (F37110-1218 does not have a pivot ring)
- Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Customers Who Bought This Also Bought