E-Grip® Powder-Free Latex Examination Gloves, Microflex®
Supplier: Ansell Healthcare
Certificates
About this item
The micro-textured satin finish of E-Grip gloves provides enhanced gripping power and excellent tactile sensitivity.
- Beaded cuffs for easy donning
- 510K approved for medical use
Gloves are nonsterile and they feature textured fingers for improved tactile sensitivity and strength.
Customers Who Bought This Also Bought