Duran Media Storage Glass Bottles, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Screw-cap glass media bottles are stable and chemically resistant.
- Chemically resistant
- Plastic pouring ring
- Interchangeable caps and rings
They are complete with a plastic pouring ring which imparts drop-free working. Plastic caps and pouring rings are completely interchangeable since all bottles from 100 ml on have the same thread size. Bottle, pouring ring and cap can be sterilized (up to 140 ˚C).
Frequently Bought Together