Diversified Biotech Reagent Reservoirs
Supplier: Diversified Biotech
Certificates
About this item
Disposable reservoirs available in sterile and nonsterile formats.
- Unique trough within a trough maximizes reagent recovery
- Convenient pour-off spouts on all four corners
- Made from high impact polystyrene
- A wide base provides stability and avoids spills
- Graduated marks on inside wall
- Superior design provides stronger sidewalls
- Sterile versions are sterilized by gamma irradiation
The 25mL divider allows for pipetting with eight channels on one side and four channels or a single channel on the other; the divider minimizes reagent loss and is convenient for adding controls.
Customers Who Bought This Also Bought