National Scientific Microtube Racks
Fournisseur: Avantor
Certificats
A propos de cet article
This system offers 1.1 mL MicroTubes or TubeStrips, Micro-Cap strips, and 8 x 12 hole MicroRacks. Design compliments multichannel pipettors and robotic work stations. Alphanumeric, non-reversing lid ensures positive sample ID. Lid, with individual condensation rings above each tube, prevents cross-contamination. Uses include harvesting of hybridoma culture supernatant, CMV and HSV testing, uptake and binding assays, and EIA/RIA serial dilution procedures. All components, except MicroCap Strips, are autoclavable.
- Can be centrifuged
- Stackable
- Available radiation sterilized, and in colors
Customers Who Bought This Also Bought