Thick Chromatography Paper, Grade 222, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Ahlstrom Grade 222 is a 100% cotton medium thick chromatography paper with a smooth and uniform texture
It offers extremely high flow and is recommended for both electrophoresis of large molecules and as a blotter to complete the blotting sandwich in gel transfer applications.
Spécifications
- Catalog No:
- 47745-908
- 47745-910
- 47749-852
- 47749-854
- 47745-916
- 47745-912
- 47745-914
- Thickness:
- 0.83 mm
- 0.83 mm
- 0.83 mm
- 0.83 mm
- 0.83 mm
- 0.83 mm
- 0.83 mm
- Absorption speed:
- 220 mm / 30 min
- 220 mm / 30 min
- 220 mm / 30 min
- 220 mm / 30 min
- 220 mm / 30 min
- 220 mm / 30 min
- 220 mm / 30 min