Cimarec™ Basic Hotplate Stirrers, Thermo Scientific
Supplier: THERMO FISHER SCIENTIFIC MX
Certificates
About this item
The sleek, rugged design of the Thermo Scientific™ Cimarec basic magnetic hotplate stirrer combines performance and economy.
Reaches maximum temperature in under 8 minutes, control knobs allow precise temperature and speed control, cleans easily, spillaway design diverts spills away from internal, and the integral ring-stand holder accommodates 1.3 cm/.5" diameter support rod components. Three year warranty.
Delivery: Supplied with TFE-coated stir bar.
Specifications
- Catalog No:
- 97027-352
- 97027-358
- Model:
- 97027-352
- 97027-358
- Plate material:
- Ceramic
- Ceramic
- Overall dimensions:
- 108L×145W×127H mm (7.9×5.7×5"), Surface area: 10.8 cm²
- 184L×211W×127H mm (11.1×8.3×5"), Surface area:18.4 cm²
- Electrical:
- 120V 60 Hz, 405 W
- 120V 60 Hz, 1086 W
- Speed range:
- 100 - 2500 rpm
- 100 - 2500 rpm
- Temperature range:
- 150…538 °C
- 150…538 °C