Ward's® Single Probes Current Sensor
# de catalogue 470014-778
Fournisseur: Wards
Spécifications
- Description:Ward's Single Probes Current Sensor
- Cat. no.:470014-778
- Range:-2.5 - 2.5 amps
- Supplier No.:WSP-102-500
Spécifications
A propos de cet article
Introduce and explore electricity concepts and safely measure electrical current with the Ward's Single Probes Current Sensor.
- Explore concepts of electricity
- Two banana plugs
- Free activity guide
This sensor easily and accurately measures the current of electronic circuits using two banana plugs, included with the sensor. Sensor includes free activity guide with activities designed for use with each probe, available digitally online after purchase. Range: -2.5 to 2.5 amps; Resolution: 1 mA. Sample rate, 10,000 per second. Requires Ward's Single Probes Base Unit, available separately.
Customers Who Bought This Also Bought